Masterflex® Gold-colored IDentifier Hinge Clamp, 3" Clamp Size, 304 SS; 1/EA
Catalog # MFLX30562-55
Supplier: Avantor Fluid Handling
About this item
Easy visual identification of critical joints for review in your process.
- Constructed of gold-coloured 304 Stainless Steel
Easily distinguishable from standard stainless steel clamps from a distance, these gold-coloured sanitary clamps are identical in size and function and can be used interchangeably with standard stainless steel clamps. Clamps feature a TiN (titanium nitride) coating. Clamps are available in a variety of sizes.
- Connection type:Sanitary
- 설명:Gold-coloured Identifier Hinge Clamp, 3" Clamp Size, 304 SS; 1/EA
- Material:Stainless steel 304
- Environmentally Preferable:
- Sanitary clamp size (port 1):3"
- Warranty:1 year
- Pk:1
- Clamp type:Sanitary
Frequently Bought Together