Masterflex® Fitting, Nylon, Elbow, Hosebarb to Thread Adapter, 5/8" ID x 3/4" NPT(M); 10/PK
Catalog # MFLX30401-24
Supplier: Avantor Fluid Handling
About this item
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 5/8" ID x 3/4" NPT(M)
Barbed elbow fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- 바브 크기/튜브 내부 직경 (포트 2):5/8"
- 연결 유형:Thread to Barb
- Flow pattern:Elbow
- Thread size NPT(M) (port 1):3/4"
- 설명:Fitting, Nylon, Elbow, Hosebarb to Thread Adapter, 5/8" ID x 3/4" NPT(M); 10/PK
- Seq:08
- Environmentally Preferable:
- Model:30401-24
- Orifice dimensions:0,46875"
- Sterilisation:Ethylene oxide (ETO) only
- Warranty:1 year
- Pk:10
- Fitting material:Nylon
Frequently Bought Together