Parker Hannifin Compression Union Tee Adapter, SS, 1/4" OD (2) x 1/4" NPT(M); Each
Catalog # MFLX31406-86
Supplier: Avantor Fluid Handling
CE certificate
About this item
Get a secure seal at higher pressures.
- Quick and easy assembly without tools
Compression and threaded fittings offer a more permanent installation. Choose the size and material to suit your application needs.
- 연결 유형:Compression to Thread
- Flow pattern:T
- 설명:Compression Union Tee Adapter, SS, 1/4" OD (2)×1/4" NPT(M); Each
- Environmentally Preferable:
- Sterilisation:Autoclave, ethylene oxide (ETO), or gamma
- Warranty:LTD life time
- Pk:1
- Fitting material:Stainless Steel 316
Frequently Bought Together