Masterflex® Fitting, Brass, Straight, Hosebarb to Thread Adapter, 1/2" ID x 3/4" NPT(M); 5/PK
Catalog # MFLX30904-42
Supplier: Avantor Fluid Handling
About this item
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
- 바브 크기/튜브 내부 직경 (포트 2):½"
- 연결 유형:Thread to Barb
- Flow pattern:Straight
- Thread size NPT(M) (port 1):¾"
- 설명:Fitting, Brass, Straight, Hosebarb to Thread Adapter, ½" ID × ¾" NPT(M); 5/PK
- Seq:12
- Environmentally Preferable:
- Model:30904-42
- No. of ports:2
- Warranty:1 year
- Pk:5
- Fitting material:Brass
Frequently Bought Together