ContactUsLinkComponent
Masterflex® Pump Extension Cable, RJ-12, 10-ft (3 m) Length; 1/Ea
Masterflex® Pump Extension Cable, RJ-12, 10-ft (3 m) Length; 1/Ea
Catalog # MFLX77095-03
undefined
Masterflex® Pump Extension Cable, RJ-12, 10-ft (3 m) Length; 1/Ea
Catalog # MFLX77095-03
About this item
Make the most of your Masterflex® pump with connection cables, replacement parts, and other accessories.
  • Choose accessories and replacement parts suitable for your Masterflex® pump
  • Increase flexibility with extended cable lengths
Convenient accessories to make your job easier.
  • 설명:
    Optional extension cable, RJ12; 10 ft (3 m).
  • Environmentally Preferable:
  • Pk:
    1
Frequently Bought Together