ContactUsLinkComponent
Masterflex® Communication Cable, Pump to Pump, RS-485, 6 ft (2 m)
Masterflex® Communication Cable, Pump to Pump, RS-485, 6 ft (2 m)
Catalog # MFLX74905-74
undefined
Masterflex® Communication Cable, Pump to Pump, RS-485, 6 ft (2 m)
Catalog # MFLX74905-74
About this item
Make the most of your Masterflex® syringe pumps with these accessories.
  • Optimize performance of your Masterflex pumps
Parts and accessories for Masterflex syringe pumps.
  • 설명:
    Communication Cable, pump to pump, RS-485, 6 ft (2 m)
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Pk:
    1
Frequently Bought Together