Masterflex® 6/10 Rack for Syringe Pump, Infusion and Withdrawal Design, Touchscreen
Catalog # MFLX74905-60
Supplier: Avantor Fluid Handling
About this item
Make the most of your Masterflex® syringe pumps with these accessories.
- Optimize performance of your Masterflex pumps
Parts and accessories for Masterflex syringe pumps.
- 설명:6/10 Rack for Syringe Pump, Infusion and Withdrawal design, Touchscreen
- Environmentally Preferable:
- Model:74905-60
- Syringe size accepted:0,5 µl - 60 ml
- Warranty:1 year
- Pk:1
Frequently Bought Together