PendoTech Cable Adapter with D15 For Single Use Pressure For PMAT(12Ft/4M) 1 * 1 items
MFLX61009-45
PendoTech Cable Adapter with D15 For Single Use Pressure For PMAT(12Ft/4M) 1 * 1 items
MFLX61009-45
Frequently Bought Together
Masterflex® Adapter Fittings, Female Luer to Hose Barb, Bulkhead, Polypropylene, Avantor®
: Avantor Fluid Handling
CPC® Quick-Disconnect Fittings, Hose Barb Bodies
: Avantor Fluid Handling
1 1/2'' Sanitary Clamp 316L Ss 1 * 1 items
: Avantor Fluid Handling
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Elbow, Nylon, Avantor®
: Avantor Fluid Handling
Masterflex® Adapter Fittings, Hose Barb Tees, PVDF, Avantor®
: Avantor Fluid Handling
Masterflex ADAPTER RJ45 TO M18 F/33502-01 1 * 1 items
: Avantor Fluid Handling
Masterflex® Union Fittings, Female NPT Threaded, Straight, Avantor®
: Avantor Fluid Handling
Masterflex® Reducer Fittings, Hose Barb, Straight (Nylon), Avantor®
: Avantor Fluid Handling
John Guest acetal push-to-connect reducer swivel adapter
: Avantor Fluid Handling
Masterflex® Reducer Fittings, Male NPT Threaded, Straight, Avantor®
: Avantor Fluid Handling
HOSE BARB IN-LINE 1/8 25/PK 1 * 25 items
: Avantor Fluid Handling
CPC® ChemQuik® Quick-Disconnect Fittings, Panel-Mount Coupling Bodies
: Avantor Fluid Handling
COVER SS K25.2 1 * 1 items
: Avantor Fluid Handling
Venture Tech ELBOW PFA 3/8''FL X 1/4''MNPT 1 * 1 items
: Avantor Fluid Handling
CPC® Plastic Quick-Disconnect Fittings, Hose Barb Bodies
: Avantor Fluid Handling
Masterflex® Barbed Medium Flow Female Quick-Disconnect Fittings, Avantor®
: Avantor Fluid Handling
Masterflex® Union Fittings, Hose Barb, Straight, Nylon, Avantor®
: Avantor Fluid Handling
Masterflex® Cleanroom 1" Sanitary Flange to Hose Barb, Avantor®
: Avantor Fluid Handling