Masterflex® Fitting, Polypropylene, Elbow, Hose Barb Unions, 1/16" ID; 10/PK
Catalog # MFLX50611-97
Supplier: Avantor Fluid Handling
CE certificate
About this item
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 1/16" ID
Create 90° bends in flexible tubing—without the kinks!
- 바브 크기/튜브 내부 직경 (포트 1):¹⁄₁₆"
- 바브 크기/튜브 내부 직경 (포트 2):¹⁄₁₆"
- 연결 유형:Barbed Union
- Flow pattern:Elbow
- 설명:Fitting, Polypropylene, Elbow, Hose Barb Unions, ¹⁄₁₆" ID; 10/PK
- Seq:28
- Environmentally Preferable:
- Model:50611-97
- Pk:10
- Fitting material:Polypropylene
Frequently Bought Together