Masterflex® Fitting, Clear Polypropylene, Straight, Hose Barb to Threaded Adapter, 1/8" ID x 1/4"-28 UNF(M); 25/PK
Catalog # MFLX31501-54
Supplier: Avantor Fluid Handling
About this item
No tools needed for assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- 바브 크기/튜브 내부 직경 (포트 2):1/8"
- 연결 유형:Thread to Barb
- Flow pattern:Straight
- 설명:Fitting, Clear Polypropylene, Straight, Hose Barb to Threaded Adapter, 1/8" ID x 1/4"-28 UNF(M); 25/PK
- Seq:08
- Environmentally Preferable:
- Model:31501-54
- Orifice dimensions:1,588 mm (0,0625")
- Sterilisation:Autoclave or ethylene oxide (ETO)
- Pk:25
- Fitting material:Polypropylene
- Thread size UNF(M) (port 2):1/4-28"
Frequently Bought Together