Masterflex® Fitting, HDPE, Tee, Hosebarb to Thread Adapter, 3/8" ID x 3/8" ID x 1/4" NTP(M); 10/PK
Catalog # MFLX30624-06
Supplier: Avantor Fluid Handling
CE certificate
About this item
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 3/8" ID x 3/8" ID x 1/4" NTP(M)
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
- 바브 크기/튜브 내부 직경 (포트 2):3/8"
- 연결 유형:Thread to Barb
- Flow pattern:T
- 설명:Fitting, HDPE, Tee, Hosebarb to Thread Adapter, 3/8" ID x 3/8" ID x 1/4" NTP(M); 10/PK
- Seq:08
- Environmentally Preferable:
- Model:30624-06
- Orifice dimensions:0,25"
- Sterilisation:Ethylene oxide (ETO) only
- Pk:10
- Fitting material:HDPE
Frequently Bought Together