Masterflex® Ismatec® Cartridge Holder
Catalog # MFLX07618-73
Supplier: Avantor Fluid Handling
About this item
Choose new cassettes based upon material of construction, tubing type and pressure capabilities.
- Material of construction standard POM-C or optional PVDF
There are two different designs of cartridge. The Click-N-Go design allows for tubing to be used in systems that have up to 1 bar of back pressure exerted on the tubing. The Pressure Lever design allows for system pressures up to 1.5 bar.
- 설명:Cartridge holder
- Seq:01
- Environmentally Preferable:
- Warranty:1 year
- Pk:2
Frequently Bought Together