Masterflex® L/S® Standard Pump Head, Mounting Hardware, Mounts Two L/S 36 Pump Head, Stainless Steel; 2/Set
Catalog # MFLX07036-02
Supplier: Avantor Fluid Handling
About this item
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
- Suitable for use with L/S® Standard Pump Heads 07036-30 and -31
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- No. of channels:2
- Pump series:L/S
- 설명:Standard Pump Head, Mounting Hardware, Mounts Two L/S® 36 Pump Head, Stainless Steel; 2/Set
- Seq:34
- Environmentally Preferable:
- Model:07036-02
- Open-head sensor:No
- Tubing sizes accepted:L/S 36
- Warranty:1 year
- Pk:1
Frequently Bought Together