Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
L/S® Pump head mounting hardware, Standard and Easy-Load®, four pump heads, stainless steel; 2/set, 2 years warranty, Stainless steel, Masterflex L/S® Easy-Load® Pump Head, L/S
MFLX07013-09
Easy stack multiple pump heads on your Masterflex® L/S® drive.
- Constructed of durable stainless steel
- See drive specifications for recommended number of heads
Stack multiple pump heads or replacement single pump head mounting hardware. Choose mounting hardware based on the Masterflex® L/S® pump head that you have. Compatible with the Masterflex L/S series pumps.
- 설명:L/S® Pump head mounting hardware, Standard and Easy-Load®, four pump heads, stainless steel; 2/set, 2 years warranty
- Material:Stainless steel
- Seq:04
- Compatible with:Masterflex L/S® Easy-Load® Pump Head
- Pump series:L/S
- Cat. No.:MFLX07013-09