Masterflex® Fitting, Polypropylene, Tee, Hose Barb Union, 1/2" ID, 100/pk
Catalog # MFLX40640-32
Supplier: Avantor Fluid Handling
CE certificate
About this item
No tools needed for assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 1/2" ID
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- 바브 크기/튜브 내부 직경 (포트 1):½"
- 바브 크기/튜브 내부 직경 (포트 2):½"
- 연결 유형:Barbed Union
- Flow pattern:T
- 설명:Fitting, Polypropylene, Tee, Hose Barb Union, ½" ID, 100/pk
- Seq:36
- Environmentally Preferable:
- Model:40640-32
- Orifice dimensions:0,34375"
- Sterilisation:Ethylene oxide (ETO) or gamma
- Warranty:Out of box
- Pk:100
- Barb size/tubing int.Ø (port 3):½"
- Fitting material:Polypropylene
Frequently Bought Together