₩0
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Korea, Republic of
ContactUsLinkComponent
100-1000µl Natural Long pipette extremely-low retention Bulk Non-sterilized Dnase/Rnase-free Non-pyrogenic 1 * 10.000 items
612-6801
:
VWR Collection
100-1000µl Natural Long pipette extremely-low retention Bulk Non-sterilized Dnase/Rnase-free Non-pyrogenic 1 * 10.000 items
612-6801
:
VWR Collection
Frequently Bought Together
Serological 피펫, standard
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer pipette, 고성능
: VWR Collection
니트릴 롱장갑
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, 두꺼운 니트릴 장갑
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
고급 화장지
: VWR Collection