건너뛰기 헤더
건너뛰기 메인 콘텐츠
건너뛰기 푸터
고급 검색
고급 검색
SDS
인증서
제조업체 데이터
0
₩0
Order Entry
로그인
/
등록
Products
Services
Digital Solutions
Applications
Support
About
주문 입력
주문 상태
귀하의 계정
장바구니
Korea, Republic of
주문 입력
주문 상태
귀하의 계정
장바구니
ContactUsLinkComponent
300µl Natural extremely-low retention Non-sterilized Dnase/Rnase-free Non-pyrogenic 1 * 10.000 items
612-6795
:
VWR Collection
300µl Natural extremely-low retention Non-sterilized Dnase/Rnase-free Non-pyrogenic 1 * 10.000 items
612-6795
:
VWR Collection
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Serological 피펫, standard
: VWR Collection
VWR®, 두꺼운 니트릴 장갑
: VWR Collection
고급 화장지
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR®, Transfer pipette, 고성능
: VWR Collection
니트릴 롱장갑
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals