Polysorbate 80 NF, Multi-Compendial, J.T.Baker®
Glass bottle, 1×1 L, 1 L
공급사: Avantor
Jump to:
Product Details & Documents
동의어: Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate
- CAS Number:9005-65-6
- MDL 번호:MFCD00082107
- 용융점:100 °C
- :Management of Change (MOC) category = R
- 인화점:148,0 °C
- 보관 온도:Ambient
- EINECS:500-019-9
- UN:0000
Documents
사양
사양 테스트 결과
- NF - Acid Value:≤ 2.0
- NF - Hydroxyl Value:65 - 80
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Residue on Ignition:≤ 0.25 %
- NF - Saponification Value:45 - 55
- NF - Specific Gravity at 25°C:1.06 - 1.09
- NF - Viscosity at 25°C, cSt:300 - 500
- NF - Water (H₂O):≤ 3.0 %
- NF - Peroxide value, meq/kg:≤ 10
- NF - Ethylene Oxide:≤ 1 ppm
- NF - Dioxane:≤ 10 ppm
- NF - Composition of Fatty Acids - Myristic Acid:≤ 5.0 %
- NF - Composition of Fatty Acids - Palmitic Acid:≤ 16.0 %
- NF - Composition of Fatty Acids - Palmitoleic Acid:≤ 8.0 %
- NF - Composition of Fatty Acids - Stearic Acid:≤ 6.0 %
- NF - Composition of Fatty Acids - Oleic Acid:≥ 58.0 %
- NF - Composition of Fatty Acids - Linoleic Acid:≤ 18.0 %
- NF - Composition of Fatty Acids - Linolenic Acid:≤ 4.0 %
- EP - Acid Value:≤ 2.0
- EP - Total Ash:≤ 0.25 %
- EP - Hydroxyl Value:65 - 80
- EP - Identification A:Passes Test
- EP - Identification D:Passes Test
- EP - Peroxide Value:≤ 10.0
- EP - Ethylene Oxide:≤ 1 ppm
- EP - Dioxan:≤ 10 ppm
- EP - Saponification Value:45 - 55
- EP - Water (H₂O):≤ 3.0 %
- EP/BP - Composition of Fatty Acids - Myristic Acid:≤ 5.0 %
- EP/BP - Composition of Fatty Acids - Palmitic Acid:≤ 16.0 %
- EP/BP - Composition of Fatty Acids - Palmitoleic Acid:≤ 8.0 %
- EP/BP - Composition of Fatty Acids - Stearic Acid:≤ 6.0 %
- EP/BP - Composition of Fatty Acids - Oleic Acid:≥ 58.0 %
- EP/BP - Composition of Fatty Acids - Linoleic Acid:≤ 18.0 %
- EP/BP - Composition of Fatty Acids - Linolenic Acid:≤ 4.0 %
- Appearance:Passes Test
- JP - Acid Value:≤ 2.0
- JP - Composition of Fatty Acids - Myristic Acid:≤ 5.0 %
- JP - Composition of Fatty Acids - Palmitic Acid:≤ 16.0 %
- JP - Composition of Fatty Acids - Palmitoleic Acid:≤ 8.0 %
- JP - Composition of Fatty Acids - Stearic Acid:≤ 6.0 %
- JP - Composition of Fatty Acids - Oleic Acid:≥ 58.0 %
- JP - Composition of Fatty Acids - Linoleic Acid:≤ 18.0 %
- JP - Composition of Fatty Acids - Linolenic Acid:≤ 4.0 %
- JP - Dioxane:≤ 10 ppm
- JP - Ethylene Oxide:≤ 1 ppm
- JP - Heavy Metals (as Pb):≤ 20 ppm
- JP - Hydroxyl Value, meq KOH/g:65 - 80
- JP - Identification:Passes Test
- JP - Peroxide Value:≤ 10.0
- JP - Residue on Ignition:≤ 0.25 %
- JP - Water (H₂O):≤ 3.0 %
- JP - Saponification Value:45 - 55
- Microbiological - Total Aerobic Microbial Count (cfu/g):≤ 100
- Microbiological - Total Yeast and Mold Count (cfu/g):≤ 50
- Bacterial endotoxins, EU/mL:≤ 50
- Ethylene glycol:≤ 0.10 %
- Diethylene glycol:≤ 0.10 %
사양
- 포장 유형:Glass bottle
- Pk:1 L
- Pk Info:1×1 L
사양
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



