Orders LinkContactUsLinkComponent
J.T.Baker®, Cycle-Tap Sampler
J.T.Baker®, Cycle-Tap Sampler
Cycle-tap sampler, hexanes, high purity, Yellow
  9815-06
 :   Avantor
J.T.Baker®, Cycle-Tap Sampler
  9815-06
 :  Avantor
J.T.Baker®, Cycle-Tap Sampler
J.T.Baker®, Cycle-Tap Sampler
Cycle-tap sampler, hexanes, high purity, Yellow
1 * 1 items
Cycle-Tap sampler is a device for sampling small quantities of solvent.
Designed to eliminate solvent exposure to the atmosphere when used with a Luer lock syringe.
  • Colour:
    Yellow
  • 설명:
    Cycle-tap sampler, hexanes, high purity
  • Seq:
    06