J.T.Baker®, Cycle-Tap Sampler
Cycle-tap sampler, hexanes, high purity, Yellow
9815-06
: Avantor
Cycle-Tap sampler is a device for sampling small quantities of solvent.
Designed to eliminate solvent exposure to the atmosphere when used with a Luer lock syringe.
- Colour:Yellow
- 설명:Cycle-tap sampler, hexanes, high purity
- Seq:06