Orders LinkContactUsLinkComponent
J.T.Baker®, Cycle-Tap Sampler
J.T.Baker®, Cycle-Tap Sampler
Cycle-tap sampler, DCM, high purity, low water, Green
Catalog # 9815-04
Supplier:   Avantor
J.T.Baker®, Cycle-Tap Sampler
Catalog # 9815-04
Supplier:  Avantor
J.T.Baker®, Cycle-Tap Sampler
J.T.Baker®, Cycle-Tap Sampler
Cycle-tap sampler, DCM, high purity, low water, Green
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Cycle-Tap sampler is a device for sampling small quantities of solvent.
Designed to eliminate solvent exposure to the atmosphere when used with a Luer lock syringe.
Specifications
  • Colour:
    Green
  • 설명:
    Cycle-tap sampler, DCM, high purity, low water
  • Seq:
    04