J.T.Baker®, Cycle-Tap Sampler
Cycle-tap sampler, methanol, high purity, low water, Orange
제품 번호 9815-03
공급사: Avantor
Jump to:
Product Details & Documents
Cycle-Tap sampler is a device for sampling small quantities of solvent.
Designed to eliminate solvent exposure to the atmosphere when used with a Luer lock syringe.
Specifications
- Colour:Orange
- 설명:Cycle-tap sampler, methanol, high purity, low water
- Seq:03