J.T.Baker®, Cycle-Tap Sampler
Cycle-tap sampler, THF, high purity, low and ultra low water, Blue
9815-01
: Avantor
Cycle-Tap sampler is a device for sampling small quantities of solvent.
Designed to eliminate solvent exposure to the atmosphere when used with a Luer lock syringe.
- Colour:Blue
- 설명:Cycle-tap sampler, THF, high purity, low and ultra low water
- Seq:01