Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
₩0
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Korea, Republic of
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Sample tested as pre-ship sample. Sample required for receipt of finished goods for Identification testing. 1 * 50 lb
Catalog # MACR7550-32
Supplier:
MACRON AVANTOR BRAND
Certificates
Expand image
Sample tested as pre-ship sample. Sample required for receipt of finished goods for Identification testing. 1 * 50 lb
Catalog # MACR7550-32
Supplier:
MACRON AVANTOR BRAND
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Product specification
spec_MACR7550-32
Documentation
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Serological 피펫, standard
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, 두꺼운 니트릴 장갑
Supplier: VWR Collection
고급 화장지
Supplier: VWR Collection
VWR®, Transfer pipette, 고성능
Supplier: VWR Collection
니트릴 롱장갑
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals