96 WELL PLATE SW 2ML WITH V-BOTTOM PP 1 * 60 items
Catalog # RITT43001-0504
Supplier: RITTER
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
고급 화장지
Supplier: VWR Collection
VWR®, 두꺼운 니트릴 장갑
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
니트릴 롱장갑
Supplier: VWR Collection
VWR®, Transfer pipette, 고성능
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Serological 피펫, standard
Supplier: VWR Collection