주문 입력
Korea, Republic of
Orders LinkContactUsLinkComponent
 

VWR®, Pipette Controller, Classic

Orange
  612-6658
VWR®, Pipette Controller, Classic
  612-6658
VWR®, Pipette Controller, Classic
VWR®, Pipette Controller, Classic

Orange

 
 
   
1 * 1 items
 

Pipette controller for glass and plastic pipettes from 1 to 100 ml.

  • Speed of suction and dispensing controlled through the finger triggers
  • Fully charged has approximately 8 h of continuous use available
  • Low battery light illuminates when 1 hour's power remains

Offers a unique combination of features to give precision fingertip control in a classic streamlined design, suitable for users with large or small hands. Gravity dispense mode is designed for use with 'To Deliver' (TD) pipettes.

 
  • Colour code:
    Orange
  • Seq:
    03