클린룸 작업복, VWR® Maximum, SF
White, S
제품 번호 113-9161
공급사: VWR Collection
Jump to:
Product Details & Documents
VWR® Maximum SF overalls are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and coated with breathable polyethylene film, these garments have been designed to optimise barrier protection, cleanliness and comfort properties.
- Available as a standard overall with collar, either elasticated ankles or non slip boot covers, or hooded overall with fluid-impervious boot covers
- Inset sleeves, zip closure, elasticated cuffs
- Superior fluid and particulate barrier
- Lowest level of particle shedding: Level 1 Helmke drum classification
- Light chemical splash resistance
- Improved breathability to ensure user comfort
- Soft, cloth-like fabric with maximum tear strength
Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
사양
- 의류 사이즈:S
- Colour:White
- Seq:16
- Cat. No.:113-9161
- Model:SF
- Garment application:Cleanroom|Laboratory
- Bacterial filtration efficiency:99,9%