Orders LinkContactUsLinkComponent
VWR® SuperSlips™, Micro Cover Glasses, Rectangular
VWR® SuperSlips™, Micro Cover Glasses, Rectangular
24×50 mm, 0,13 - 0,17 mm
제품 번호 VWRU48404-453
공급사:   VWR Collection
VWR® SuperSlips™, Micro Cover Glasses, Rectangular
제품 번호 VWRU48404-453
공급사:  VWR Collection
VWR® SuperSlips™, Micro Cover Glasses, Rectangular
VWR® SuperSlips™, Micro Cover Glasses, Rectangular
24×50 mm, 0,13 - 0,17 mm
Jump to:
Price
₩27,938
Log in 재고 확인
1 * 1 items
Product Details & Documents
These cover glasses are corrosion-resistant and uniformly thin and flat.
  • Suitable for humid environments
  • Resistant to corrosion
  • Processing techniques result in superior cleanliness
SuperSlip coverslips combine specialised production methods with moisture-resistant packaging to prevent sticking.
사양
  • 길이×너비:
    24×50 mm
  • Seq:
    01
  • 두께:
    0,13 - 0,17 mm