주문 입력
Korea, Republic of
Orders LinkContactUsLinkComponent
Rankem®, Specific Gravity Bottles, Class A

Rankem®, Specific Gravity Bottles, Class A

Specific gravity bottle, 25 ml
제품 번호 RANKBSG-PST-0025-A
공급사:   RANKEM
Rankem®, Specific Gravity Bottles, Class A
제품 번호 RANKBSG-PST-0025-A
공급사:  RANKEM
Rankem®, Specific Gravity Bottles, Class A

Specific gravity bottle, 25 ml

Jump to:
Price
가격 문의
Log in 재고 확인
1 * 5 items
Product Details & Documents

Specific Gravity Bottle Class A with Ground in Thermometer range 0 to 50 °C with work Certificate (Pyknometer). Suitable for determining the density with built in thermometer.

사양
  • 설명:
    Specific gravity bottle, 25 ml
  • Seq:
    01