VWR® Basic, Beard Covers
White
Supplier: VWR Collection
Jump to:
Product Details & Documents
This disposable beard cover is made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Free from natural rubber latex
- Available in white and blue
Weight: 10 g/m²
Packaging: 100 pieces/bag, 10 bags/case.
Documents
CoC VWR® Basic, Beard Covers coc_VWRI113-8902_05120239 coc_VWRI113-8902_19120256 COC_VWRI113-8902_22920201 coc_VWRI113-8902_24120241
More
Food contact certificate F
Additional Product Details
TDS VWR® Basic Beard Cover
Datasheet
VWR® Non Regulated Disposables for Production
Product Literature
Specifications
- Colour:White
- Cat. No.:113-8902
- Weight:10 g/m²