VWR®, IOS W-Plan Objectives
IOS W-Plan objective (oil), 100×/1,25 (i)
제품 번호 630-2955
공급사: VWR Collection
Jump to:
Product Details & Documents
These high contrast objectives are ideal for routine observations in laboratories.
- Ensure field flatness up to F.N. 22
- Based on infinity-corrected optical system
- 100× is also available with iris for darkfield
사양
- 확대:100×/1,25 (i)
- 설명:IOS W-Plan objective (oil)
- Seq:156