Orders LinkContactUsLinkComponent
VWR®, Spatulas
VWR®, Spatulas
Standard blade, —, 305
제품 번호 VWRU82027-468
공급사:   VWR Collection
VWR®, Spatulas
제품 번호 VWRU82027-468
공급사:  VWR Collection
VWR®, Spatulas
VWR®, Spatulas
Standard blade, —, 305
Jump to:
Price
₩85,538
Log in 재고 확인
1 * 1 items
Product Details & Documents
Stainless steel spatulas with wooden handle. Economical tools for handling, transferring, blending, or collecting chemical and biological samples.
  • Sturdy and durable
  • Stainless steel construction
  • Multiple sizes allow for varied laboratory applications
사양
  • 설명:
    Standard blade
  • 길이:
  • Seq:
    06
  • Blade length:
    305