VWR®, Spatulas
Standard blade, —, 127
제품 번호 VWRU82027-460
공급사: VWR Collection
Jump to:
Product Details & Documents
Stainless steel spatulas with wooden handle. Economical tools for handling, transferring, blending, or collecting chemical and biological samples.
- Sturdy and durable
- Stainless steel construction
- Multiple sizes allow for varied laboratory applications
사양
- 설명:Standard blade
- 길이:—
- Seq:02
- Blade length:127