주문 입력
Korea, Republic of
Orders LinkContactUsLinkComponent
VWR® SuperSlips™, Micro Cover Glasses, Rectangular

VWR® SuperSlips™, Micro Cover Glasses, Rectangular

24×60 mm, 0,13 - 0,17 mm
제품 번호 VWRU48404-455
공급사:   VWR Collection
VWR® SuperSlips™, Micro Cover Glasses, Rectangular
제품 번호 VWRU48404-455
공급사:  VWR Collection
VWR® SuperSlips™, Micro Cover Glasses, Rectangular
VWR® SuperSlips™, Micro Cover Glasses, Rectangular

24×60 mm, 0,13 - 0,17 mm

Jump to:
Price
₩27,751
Log in 재고 확인
1 * 1 oz
Product Details & Documents

These cover glasses are corrosion-resistant and uniformly thin and flat.

  • Suitable for humid environments
  • Resistant to corrosion
  • Processing techniques result in superior cleanliness

SuperSlip coverslips combine specialised production methods with moisture-resistant packaging to prevent sticking.

사양
  • 길이×너비:
    24×60 mm
  • Seq:
    02
  • 두께:
    0,13 - 0,17 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.