VWR®, 시약 보틀, 좁은 목, 표준 원형 마개
1000, 29/32, 105 mm, 225 mm
공급사: VWR Collection
Jump to:
Product Details & Documents
Borosilicate 3.3 glass according to ISO 4796.
- High resistance to chemicals and thermal shock
- Available in capacities from 10 to 2000 ml
- Hollow glass stopper and ground joint
These reagent bottles have a precise ground neck to ensure a water- and air-tight connection between the bottle and the glass stopper. All components are made from borosilicate 3.3 glass and are, therefore, ideal for the storage of aggressive and hot media.
사양
- 외부 직경:105 mm
- NS:29/32
- 용량:1000
- 높이:225 mm
- Seq:07



