VWR®, 시약 보틀, 좁은 목, 표준 원형 마개
50, 14/23, 42 mm, 78 mm
공급사: VWR Collection
Jump to:
Product Details & Documents
Borosilicate 3.3 glass according to ISO 4796.
- High resistance to chemicals and thermal shock
- Available in capacities from 10 to 2000 ml
- Hollow glass stopper and ground joint
These reagent bottles have a precise ground neck to ensure a water- and air-tight connection between the bottle and the glass stopper. All components are made from borosilicate 3.3 glass and are, therefore, ideal for the storage of aggressive and hot media.
사양
- 외부 직경:42 mm
- NS:14/23
- 용량:50
- 높이:78 mm
- Seq:03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



