주문 입력
Korea, Republic of
Orders LinkContactUsLinkComponent
 

VWR®, Bottle assemblies, PC, 멸균, 일회용

2000, 53B, Port 1: PP Tube Plug
Port 2: 50 mm PTFE Vent Filter
Port 3: PP Tube Plug, Port 1: 12"
Port 2: 3"
Port 3: 12", C-Flex, Port 1: 3/8 x 1/2"
Port 2: 1/4 x 3/8"
Port 3: 3/8 x 1/2"
제품 번호 734-2856
공급사:   VWR Collection
VWR®, Bottle assemblies, PC, 멸균, 일회용
제품 번호 734-2856
공급사:  VWR Collection
VWR®, Bottle assemblies, PC, 멸균, 일회용
VWR®, Bottle assemblies, PC, 멸균, 일회용

2000, 53B, Port 1: PP Tube Plug
Port 2: 50 mm PTFE Vent Filter
Port 3: PP Tube Plug, Port 1: 12"
Port 2: 3"
Port 3: 12", C-Flex, Port 1: 3/8 x 1/2"
Port 2: 1/4 x 3/8"
Port 3: 3/8 x 1/2"

Jump to:
Price
가격 문의
Log in 재고 확인
1 * 2 items
Product Details & Documents

Sterile polycarbonate containers, with weldable tubing, provide the maximum amount of flexibility and reliability. These vessels are assembled in an ISO Class 7 cleanroom and are manufactured from USP Class VI materials. They comes in a comprehensive size selection with tubing that is completely weldable/sealable.

  • Sterile bottles come individually bagged and process ready
  • Multiple bottle sizes allow for process-specific flexibility
  • When used with sterile VWR® tube sets, custom possibilities are endless
  • Gamma irradiated to Sterility Assurance Level (SAL) 10-6

Assemblies have been designed with a broad spectrum of applications in mind, including life sciences and general lab use, environmental assays, also production and cleanroom applications. All product contact materials are animal-free or processed in accordance to EMEA/410/01. The assemblies are able to withstand temperatures from –20 to +65 °C without losing integrity. All assemblies are individually double-bagged with product label located on the inner bag. Gamma irradiated to Sterility Assurance Level (SAL) 10-6.

 : *The product(s) on this page are outside the scope of the EU Directive 2019/904.

사양
  • 캡 크기:
    53B
  • 용량:
    2000
  • Seq:
    06
  • Tubing connection:
    Port 1: PP Tube Plug
    Port 2: 50 mm PTFE Vent Filter
    Port 3: PP Tube Plug
  • Tubing length:
    Port 1: 12"
    Port 2: 3"
    Port 3: 12"
  • Tubing size:
    Port 1: 3/8 x 1/2"
    Port 2: 1/4 x 3/8"
    Port 3: 3/8 x 1/2"
  • Tubing material:
    C-Flex