주문 입력
Korea, Republic of
Orders LinkContactUsLinkComponent
VWR®, Bottle assemblies, PC, 멸균, 일회용

VWR®, Bottle assemblies, PC, 멸균, 일회용

1000, 38-430, Port 1: PP Tube Plug
Port 2: 25 mm Hydrophobic PE Vent Filter, Port 1: 24"
Port 2: 3", C-Flex, Port 1: 1/8 x 1/4"
Port 2: 1/8 x 1/4"
제품 번호 734-2855
공급사:   VWR Collection
VWR®, Bottle assemblies, PC, 멸균, 일회용
제품 번호 734-2855
공급사:  VWR Collection
VWR®, Bottle assemblies, PC, 멸균, 일회용
VWR®, Bottle assemblies, PC, 멸균, 일회용

1000, 38-430, Port 1: PP Tube Plug
Port 2: 25 mm Hydrophobic PE Vent Filter, Port 1: 24"
Port 2: 3", C-Flex, Port 1: 1/8 x 1/4"
Port 2: 1/8 x 1/4"

Jump to:
Price
가격 문의
Log in 재고 확인
1 * 3 items
Product Details & Documents

Sterile polycarbonate containers, with weldable tubing, provide the maximum amount of flexibility and reliability. These vessels are assembled in an ISO Class 7 cleanroom and are manufactured from USP Class VI materials. They comes in a comprehensive size selection with tubing that is completely weldable/sealable.

  • Sterile bottles come individually bagged and process ready
  • Multiple bottle sizes allow for process-specific flexibility
  • When used with sterile VWR® tube sets, custom possibilities are endless
  • Gamma irradiated to Sterility Assurance Level (SAL) 10-6

Assemblies have been designed with a broad spectrum of applications in mind, including life sciences and general lab use, environmental assays, also production and cleanroom applications. All product contact materials are animal-free or processed in accordance to EMEA/410/01. The assemblies are able to withstand temperatures from –20 to +65 °C without losing integrity. All assemblies are individually double-bagged with product label located on the inner bag. Gamma irradiated to Sterility Assurance Level (SAL) 10-6.

주의: *The product(s) on this page are outside the scope of the EU Directive 2019/904.

사양
  • 캡 크기:
    38-430
  • 용량:
    1000
  • Seq:
    04
  • Tubing connection:
    Port 1: PP Tube Plug
    Port 2: 25 mm Hydrophobic PE Vent Filter
  • Tubing length:
    Port 1: 24"
    Port 2: 3"
  • Tubing size:
    Port 1: 1/8 x 1/4"
    Port 2: 1/8 x 1/4"
  • Tubing material:
    C-Flex
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.