VWR®, Bottle assemblies, PC, 멸균, 일회용
Port 2: 25 mm Hydrophobic PE Vent Filter, Port 1: 24"
Port 2: 3", C-Flex, Port 1: 1/8 x 1/4"
Port 2: 1/8 x 1/4"
Sterile polycarbonate containers, with weldable tubing, provide the maximum amount of flexibility and reliability. These vessels are assembled in an ISO Class 7 cleanroom and are manufactured from USP Class VI materials. They comes in a comprehensive size selection with tubing that is completely weldable/sealable.
- Sterile bottles come individually bagged and process ready
- Multiple bottle sizes allow for process-specific flexibility
- When used with sterile VWR® tube sets, custom possibilities are endless
- Gamma irradiated to Sterility Assurance Level (SAL) 10-6
Assemblies have been designed with a broad spectrum of applications in mind, including life sciences and general lab use, environmental assays, also production and cleanroom applications. All product contact materials are animal-free or processed in accordance to EMEA/410/01. The assemblies are able to withstand temperatures from –20 to +65 °C without losing integrity. All assemblies are individually double-bagged with product label located on the inner bag. Gamma irradiated to Sterility Assurance Level (SAL) 10-6.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904.
- 캡 크기:38-430
- 용량:125
- Seq:01
- Tubing connection:Port 1: PP Tube Plug
Port 2: 25 mm Hydrophobic PE Vent Filter - Tubing length:Port 1: 24"
Port 2: 3" - Tubing size:Port 1: 1/8 x 1/4"
Port 2: 1/8 x 1/4" - Tubing material:C-Flex



