VWR®, Storage Boxes
120×90×50 mm
제품 번호 216-4334
공급사: VWR Collection
Jump to:
Product Details & Documents
These boxes are an ideal choice for laboratories due to their high transparency and excellent durability.
- Stackable, transparent boxes
- For storage and presentation
- Six different sizes
- PS, clear
Documents
사양
- Seq:03
- W×D×H:120×90×50 mm