VWR®, Staining Plates
12, 112×92 mm
제품 번호 VWRU89038-188
공급사: VWR Collection
Jump to:
Product Details & Documents
Porcelain staining plate, glased (except for bottom surface).
사양
- 웰 수:12
- 길이×너비:112×92 mm
- Seq:02