Orders LinkContactUsLinkComponent
VWR®, Staining Plates
VWR®, Staining Plates
12, 112×92 mm
제품 번호 VWRU89038-188
공급사:   VWR Collection
VWR®, Staining Plates
제품 번호 VWRU89038-188
공급사:  VWR Collection
VWR®, Staining Plates
VWR®, Staining Plates
12, 112×92 mm
Jump to:
Price
₩55,615
Log in 재고 확인
1 * 1 Each
Product Details & Documents
Porcelain staining plate, glased (except for bottom surface).
사양
  • 웰 수:
    12
  • 길이×너비:
    112×92 mm
  • Seq:
    02