주문 입력
Korea, Republic of
Orders LinkContactUsLinkComponent
 

VWR®, Micro cover glasses, rectangular

22×60 mm
Catalog # VWRU48393-070
Supplier:   VWR Collection
VWR®, Micro cover glasses, rectangular
Catalog # VWRU48393-070
Supplier:  VWR Collection
VWR®, Micro cover glasses, rectangular
VWR®, Micro cover glasses, rectangular

22×60 mm

Price
₩26,215
Log in for availability
1 * 28,35 g
Product Details & Documents

These micro cover glasses are made of glass, and each set of numbered slides is uniform in size and thickness.

The No. 1 slides are 0,13 to 0,17 mm in thickness. The No. 1.5 slides are 0,16 to 0,19 mm in thickness.

Specifications
  • 길이×너비:
    22×60 mm
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.