주문 입력
Korea, Republic of
Orders LinkContactUsLinkComponent
 

VWR®, 흡수제 - 화학 물질 유출을 위한 매트,롤 및 양말

Absorbent pads for chemicals, green, 136, 510×410 mm
  121-0139
VWR®, 흡수제 - 화학 물질 유출을 위한 매트,롤 및 양말
  121-0139
VWR®, 흡수제 - 화학 물질 유출을 위한 매트,롤 및 양말
VWR®, 흡수제 - 화학 물질 유출을 위한 매트,롤 및 양말

Absorbent pads for chemicals, green, 136, 510×410 mm

 
 
   
1 * 200 items
 

VWR® absorbents and spill containment products help keep your employees and work environment safe from unexpected spills, leaks, drips or other accidents.

  • Absorbs a wide range of chemicals, including acids and caustics
  • Chemically inert: Product does not degrade or cause a dangerous chemical reaction with the adsorbed liquid
  • Green colour alerts that product is being used for hazardous liquids
  • Green colour allows user to identify and separate hazardous waste for disposal, resulting in lower disposal costs
  • Cannot be left outdoors as they will absorb water

For use with chemicals.

Colour: Green.

 
  • 용량:
    136
  • 설명:
    Absorbent pads for chemicals, green
  • 길이×너비:
    510×410 mm
Complementary Products

 

  •  
    Packed
    길이×너비
    용량
    설명
     
     
  • 121-0138
    Box
    510×410 mm
    136
    Absorbent pads for, oil, water, chemicals, grey
       
    1 * 200 items - ₩0

  • 121-0144
    Box
    1220×76 mm
    43
    Absorbent socks for, oil, water, chemicals, grey
       
    1 * 12 items - ₩0

  • 121-0141
    Dispenser box
    46000×380 mm
    57
    Absorbent roll for oil, water, chemicals, grey
       
    1 * 1 Roll - ₩0