VWR®, 흡수제 - 화학 물질 유출을 위한 매트,롤 및 양말
Absorbent roll for chemicals, green, 57, 46000×380 mm
Catalog # 121-0142
Supplier: VWR Collection
Jump to:
Product Details & Documents
VWR® absorbents and spill containment products help keep your employees and work environment safe from unexpected spills, leaks, drips or other accidents.
- Absorbs a wide range of chemicals, including acids and caustics
- Chemically inert: Product does not degrade or cause a dangerous chemical reaction with the adsorbed liquid
- Green colour alerts that product is being used for hazardous liquids
- Green colour allows user to identify and separate hazardous waste for disposal, resulting in lower disposal costs
- Cannot be left outdoors as they will absorb water
For use with chemicals.
Colour: Green.
Specifications
- 용량:57
- 설명:Absorbent roll for chemicals, green
- 길이×너비:46000×380 mm
Complementary Products
Product Information
- Catalog #설명용량Packed길이×너비AvailabilityPrice
- 121-0138Absorbent pads for, oil, water, chemicals, grey136Box510×410 mmLog in for availability1 * 200 items - ₩0
- 121-0144Absorbent socks for, oil, water, chemicals, grey43Box1220×76 mmLog in for availability1 * 12 items - ₩0
- 121-0141Absorbent roll for oil, water, chemicals, grey57Dispenser box46000×380 mmLog in for availability1 * 1 Roll - ₩0
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



