Polysorbate 80, J.T.Baker®
Glass bottle, amber, narrow mouth, 500 mL
공급사: Avantor
Jump to:
Product Details & Documents
Polyoxyethylene (20) Sorbitan Monooleate
동의어: Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate
- CAS Number:9005-65-6
- MDL 번호:MFCD00082107
- 용융점:100 °C
- 인화점:113 °C (closed cup)
- 보관 온도:Ambient
- EINECS:500-018-3
- UN:0000
Documents
Certificates
Lot 번호로 검색사양
사양 테스트 결과
- For Laboratory,Research,or Manufacturing Use:Passes Test
- Identification (by IR):Passes Test
사양
- 포장 유형:Glass bottle, amber, narrow mouth
- Pk:500 mL



