VWR®, N-Plan Objectives
N-plan objective, 40×/0,65 mm
제품 번호 630-2007
공급사: VWR Collection
Jump to:
Product Details & Documents
These objectives are ideal for use in the education field and for laboratory routine applications.
IOS N-Plan objectives feature an infinity-corrected optical system.
Documents
VWR® for Microscopy
Product Literature
사양
- 확대:40×/0,65 mm
- 설명:N-plan objective
- Seq:210