VWR®, N-Plan Objectives
N-plan objective, 10×/0,25 mm
Catalog # 630-2005
Supplier: VWR Collection
Jump to:
Product Details & Documents
These objectives are ideal for use in the education field and for laboratory routine applications.
IOS N-Plan objectives feature an infinity-corrected optical system.
Documents
VWR® for Microscopy
제품 브로셔
Specifications
- 확대:10×/0,25 mm
- 설명:N-plan objective
- Seq:208