VWR®, Cell Holders
Cell holder, 10 mm water-jacketed four-cell holder (B), for double beam only
제품 번호 634-6050
공급사: VWR Collection
Jump to:
Product Details & Documents
Documents
사양
- 설명:Cell holder, 10 mm water-jacketed four-cell holder (B), for double beam only
- Seq:17