VWR®, Cell Holders
Cell holder, four-cell holder for 10 - 50 mm square cuvette (B), for double beam only
제품 번호 634-6045
공급사: VWR Collection
Jump to:
Product Details & Documents
Documents
사양
- 설명:Cell holder, four-cell holder for 10 - 50 mm square cuvette (B), for double beam only
- Seq:19