Orders LinkContactUsLinkComponent
VWR®, Micro cover glasses, rectangular
24×60 mm
Catalog # VWRU48393-251
Supplier:   VWR Collection
VWR®, Micro cover glasses, rectangular
Catalog # VWRU48393-251
Supplier:  VWR Collection
VWR®, Micro cover glasses, rectangular
VWR®, Micro cover glasses, rectangular
24×60 mm
Price
Inquire For Price
Log in for availability
1 * 28,35 g
Product Details & Documents
These micro cover glasses are made of glass, and each set of numbered slides is uniform in size and thickness.
The No. 1 slides are 0,13 to 0,17 mm in thickness. The No. 1.5 slides are 0,16 to 0,19 mm in thickness.
Specifications
  • 길이×너비:
    24×60 mm
  • Seq:
    16