Orders LinkContactUsLinkComponent
J.T.Baker®, Cycle-Tap Sampler
Cycle-tap sampler, Brown
제품 번호 9815-05
공급사:   Avantor
J.T.Baker®, Cycle-Tap Sampler
제품 번호 9815-05
공급사:  Avantor
J.T.Baker®, Cycle-Tap Sampler
J.T.Baker®, Cycle-Tap Sampler
Cycle-tap sampler, Brown
Jump to:
Price
가격 문의
Log in 재고 확인
1 * 1 items
Product Details & Documents
Cycle-Tap sampler is a device for sampling small quantities of solvent.
Designed to eliminate solvent exposure to the atmosphere when used with a Luer lock syringe.
사양
  • Colour:
    Brown
  • 설명:
    Cycle-tap sampler
  • Seq:
    05